missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Melting Point | -30°C |
| Color | Colorless |
| Boiling Point | 174°C |
| Packaging | Amber glass bottle |
| Quantity | 100 mL |
| Formula Weight | 142.28g/mol |
| Physical Form | Liquid |
Safety and Handling
Product Identifier
- Decane
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- H304-May be fatal if swallowed and enters airways.
- H226-Flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?