missing translation for 'onlineSavingsMsg'
Learn More

n-Decyl acetate, 98%

Product Code. 11413098
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
Unit Size:
1000g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413098 250 g 250g
11478750 500 g 500g
11403098 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413098 Supplier Thermo Scientific Alfa Aesar Supplier No. A18814.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 112-17-4 | C12H24O2 | 200.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-17-4
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00026538
InChI Key NUPSHWCALHZGOV-UHFFFAOYSA-N
Synonym n-decyl acetate, acetic acid, decyl ester, n-decyl ethanoate, 1-decanol acetate, decyl ethanoate, acetate c 10, decanyl acetate, 1-decyl acetate, acetic acid n-decyl ester, decanol acetate
PubChem CID 8167
IUPAC Name decyl acetate
SMILES CCCCCCCCCCOC(C)=O

Specifications

Density 0.867
Boiling Point 244°C
Flash Point 125°C (257°F)
Refractive Index 1.427
Quantity 250 g
Beilstein 1762123
Formula Weight 200.32
Percent Purity 98%
Chemical Name or Material n-Decyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.