missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Decyl-d21-trimethylammonium Bromide, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Decyltrimethylammonium-D21 (decyl-D21) Bromide |
| CAS | 1515861-67-2 |
| Molecular Formula | C13H9D21BrN |
| Quantity | 0.1 g |
| Synonym | 1-Decan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-d21-aminium, N,N,N-trimethyl-, bromide (1:1) (ACI), Decyl-d21-trimethylammonium bromide, Decyltrimethylammonium-D21 (decyl-D21) bromide, Decyltrimethylammonium-d21 bromide, N,N,N-Trimethyl-1-decan-d21-a |
| InChI Formula | InChI=1 S/C13H30N.BrH/c1-5-6-7-8-9-10-11-12-13-14(2,3)4;/h5-13H2,1-4H3;1 H/q+1;/p-1/i1D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2; |
| SMILES | C(C(C(C(C(C(C(C(C([2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])([2 H])[2 H])(C([N+](C)(C)C)([2 H])[2 H])([2 H])[2 H].[Br-] |
| IUPAC Name | N,N,N-trimethyldecan-1-aminium-d21 bromide |
| Molecular Weight (g/mol) | 301.42 |
| Formula Weight | 300.29 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction