missing translation for 'onlineSavingsMsg'
Learn More

n-Decylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15465325
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465325 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15465325 Supplier Thermo Scientific Alfa Aesar Supplier No. H58155.AD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
n-Decylzinc bromide acts as a ligand.

Solubility
Reacts with water.

Notes
Incompatible with oxidizing agents, acid chlorides. Do not together with acids. Air Sensitive. Store under dry inert gas. St
TRUSTED_SUSTAINABILITY

Specifications

CAS 160516-17-6
Molecular Formula C10H21BrZn
Linear Formula CH3(CH2)9ZnBr
MDL Number MFCD07698739
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym n-decylzinc bromide, decylzinc bromide, bromozinc 1+ ,decane, bromo decyl zinc, n-decylzinc bromide 0.5 m in tetrahydrofuran, n-decylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key RTHCUVJQVVJPTJ-UHFFFAOYSA-M
SMILES CCCCCCCCC[CH2-].[Zn+]Br
IUPAC Name bromozinc(1+);decane
Molecular Weight (g/mol) 286.562
PubChem CID 14931676
Formula Weight 286.56
Chemical Name or Material n-Decylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.