missing translation for 'onlineSavingsMsg'
Learn More

N-Des[2-(2-hydroxyethoxy)ethyl] Quetiapine Dihydrochloride (1.0 mg/mL in Methanol), TRC

Product Code. 30404618
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30404618 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404618 Supplier Toronto Research Chemicals Supplier No. TRCKIT136510X1ML
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 111974-74-4
Molecular Formula C17 H17 N3 S . 2 Cl H
Quantity 10 x 1 mL
Synonym 11-(Piperazin-1-yl)dibenzo[b,f][1,4]thiazepine Dihydrochloride, Quetiapine Fumarate Imp. B (EP) as Dihydrochloride, Dibenzo[b,f][1,4]thiazepine, 11-(1-piperazinyl)-, hydrochloride (1:2), Dibenzo[b,f][1,4]thiazepine, 11-(1-piperazinyl)-, dihydrochloride (9CI), 11-(1-Piperazinyl)-dibenzo[b,f][1,4]thiazepine dihydrochloride, 11-Piperazin-1-yl-dibenzo[b,f][1,4]thiazepine dihydrochloride, 11-Piperazinodibenzo[b,f][1,4]thiazepine dihydrochloride, Norquetiapine dihydrochloride, N-Des[2-(2-hydroxyethoxy)ethyl]quetiapine dihydrochloride
InChI Formula InChI=1S/C17H17N3S.2ClH/c1-3-7-15-13(5-1)17(20-11-9-18-10-12-20)19-14-6-2-4-8-16(14)21-15;;/h1-8,18H,9-12H2;2*1H
SMILES Cl.Cl.C1CN(CCN1)C2=Nc3ccccc3Sc4ccccc24
IUPAC Name 6-piperazin-1-ylbenzo[b][1,4]benzothiazepine;dihydrochloride
Molecular Weight (g/mol) 368.32
Formula Weight 367.0677
Concentration or Composition (by Analyte or Components) 11-(Piperazin-1-yl)dibenzo[b,f][1,4]thiazepine Dihydrochloride 1 mg/mL
Chemical Name or Material 11-(Piperazin-1-yl)dibenzo[b,f][1,4]thiazepine Dihydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.