missing translation for 'onlineSavingsMsg'
Learn More

N-Deschlorobenzoyl Indomethacin, TRC

Product Code. 30003754
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003754 10 mg 10mg
30008523 1 g 1g
30011357 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003754 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD28884510MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2882-15-7
Molecular Formula C12 H13 N O3
Molecular Weight (g/mol) 219.24
Synonym (5-Methoxy-2-methyl-1H-indol-3-yl)acetic acid, 2-(5-Methoxy-2-methyl-1H-indol-3-yl)acetic acid, 2-(5-Methoxy-2-methylindol-3-yl)acetic acid, 2-Methyl-5-methoxy-3-indolylacetic acid, 2-Methyl-5-methoxyindole-3-acetic acid, 5-Methoxy-2-methyl-1H-Indole-3-acetic acid, 5-Methoxy-2-methyl-3-indolylacetic acid, 5-Methoxy-2-methylindol-3-acetic acid, 5-Methoxy-2-methylindole-3-acetic acid, De(chlorobenzoyl)indomethacin, N-Des(4-chlorobenzoyl)indomethacin, NSC 97026
IUPAC Name 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid
SMILES COc1ccc2[nH]c(C)c(CC(=O)O)c2c1

Tekniske data

Quantity 10 mg
InChI Formula InChI=1S/C12H13NO3/c1-7-9(6-12(14)15)10-5-8(16-2)3-4-11(10)13-7/h3-5,13H,6H2,1-2H3,(H,14,15)
Formula Weight 219.0895
Chemical Name or Material 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.