missing translation for 'onlineSavingsMsg'
Learn More

N-Deschlorobenzoyl Indomethacin, TRC

Product Code. 30011357
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011357 100 mg 100mg
30003754 10 mg 10mg
30008523 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011357 Supplier Toronto Research Chemicals Supplier No. TRCD288845100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2882-15-7
Molecular Formula C12 H13 N O3
Molecular Weight (g/mol) 219.24
Synonym (5-Methoxy-2-methyl-1H-indol-3-yl)acetic acid, 2-(5-Methoxy-2-methyl-1H-indol-3-yl)acetic acid, 2-(5-Methoxy-2-methylindol-3-yl)acetic acid, 2-Methyl-5-methoxy-3-indolylacetic acid, 2-Methyl-5-methoxyindole-3-acetic acid, 5-Methoxy-2-methyl-1H-Indole-3-acetic acid, 5-Methoxy-2-methyl-3-indolylacetic acid, 5-Methoxy-2-methylindol-3-acetic acid, 5-Methoxy-2-methylindole-3-acetic acid, De(chlorobenzoyl)indomethacin, N-Des(4-chlorobenzoyl)indomethacin, NSC 97026
IUPAC Name 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid
SMILES COc1ccc2[nH]c(C)c(CC(=O)O)c2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H13NO3/c1-7-9(6-12(14)15)10-5-8(16-2)3-4-11(10)13-7/h3-5,13H,6H2,1-2H3,(H,14,15)
Formula Weight 219.0895
Chemical Name or Material 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.