missing translation for 'onlineSavingsMsg'
Learn More

N-Desethyl Amodiaquine Hydrochloride, TRC

Product Code. 17953335
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953335 50 mg 50mg
17950375 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17953335 Supplier Toronto Research Chemicals Supplier No. TRCD28882650MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79049-30-2
Molecular Formula C18 H18 Cl N3 O . 2 Cl H
Molecular Weight (g/mol) 400.73
Synonym Monodesethylamodiaquine dihydrochloride, Phenol, 4-[(7-chloro-4-quinolinyl)amino]-2-[(ethylamino)methyl]-, hydrochloride (1:2), Phenol, 4-[(7-chloro-4-quinolinyl)amino]-2-[(ethylamino)methyl]-, dihydrochloride (9CI), o-Cresol, 4-(7-chloro-4-quinolylamino)-α-ethylamino-, dihydrochloride (5CI), 4-[(7-Chloro-4-quinolinyl)amino]-2-[(ethylamino)methyl]phenol dihydrochloride, Desethylamodiaquine dihydrochloride, N-Desethylamodiaquine dihydrochloride
IUPAC Name 4-[(7-chloroquinolin-4-yl)amino]-2-(ethylaminomethyl)phenol;dihydrochloride
SMILES Cl.Cl.CCNCc1cc(Nc2ccnc3cc(Cl)ccc23)ccc1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H18ClN3O.2ClH/c1-2-20-11-12-9-14(4-6-18(12)23)22-16-7-8-21-17-10-13(19)3-5-15(16)17;;/h3-10,20,23H,2,11H2,1H3,(H,21,22);2*1H
Formula Weight 399.0672
Percent Purity >95%
Chemical Name or Material N-Desethylamodiaquine dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.