missing translation for 'onlineSavingsMsg'
Learn More

N-Desethyl N-Methyl rac-Rivastigmine, TRC

Product Code. 30357333
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357333 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357333 Supplier Toronto Research Chemicals Supplier No. TRCD292300100MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R,S)-3-[1-(Dimethylamino)ethyl]phenyl dimethylcarbamate
CAS 25081-93-0
Molecular Formula C13 H20 N2 O2
Quantity 100 mg
Synonym N,N-Dimethylcarbamic acid 3-[1-(Dimethylamino)ethyl]phenyl ester, Dimethylcarbamic acid 3-[1-(Dimethylamino)ethyl]phenyl ester, Dimethylcarbamic acid m-[1-(Dimethylamino)ethyl]phenyl ester, m-[1-(Dimethylamino)ethyl]phenol dimethylcarbamate, Carbamic acid, dimethyl-, 3-[1-(dimethylamino)ethyl]phenyl ester (9CI), Carbamic acid, dimethyl-, m-[1-(dimethylamino)ethyl]phenyl ester (8CI), 3-[1-(Dimethylamino)ethyl]phenyl N,N-dimethylcarbamate, Phenol, m-[1-(dimethylamino)ethyl]-, dimethylcarbamate (ester) (8CI), 3-[(1RS)-1-(Dimethylamino)ethyl]phenyl Dimethylcarbamate, N-Desethyl N-Methyl rac-Rivastigmine
InChI Formula InChI=1S/C13H20N2O2/c1-10(14(2)3)11-7-6-8-12(9-11)17-13(16)15(4)5/h6-10H,1-5H3
SMILES CC(N(C)C)c1cccc(OC(=O)N(C)C)c1
IUPAC Name [3-[1-(dimethylamino)ethyl]phenyl] N,N-dimethylcarbamate
Molecular Weight (g/mol) 236.31
Formula Weight 236.1525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.