missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Azelastine, TRC

Product Code. 30350739
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350739 5 mg 5mg
30350278 25 mg 25mg
30366585 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30350739 Supplier Toronto Research Chemicals Supplier No. TRCD2909505MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 47491-38-3
Molecular Formula C21 H22 Cl N3 O
Molecular Weight (g/mol) 367.87
Synonym 1(2H)-Phthalazinone, 4-[(4-chlorophenyl)methyl]-2-(hexahydro-1H-azepin-4-yl)-, 4-[(4-Chlorophenyl)methyl]-2-(hexahydro-1H-azepin-4-yl)-1(2H)-phthalazinone, Demethylazelastine, Desmethylazelastine, N-Desmethyl Azelastine, N-Desmethylazelastine
IUPAC Name 2-(azepan-4-yl)-4-[(4-chlorophenyl)methyl]phthalazin-1-one
SMILES Clc1ccc(CC2=NN(C3CCCNCC3)C(=O)c4ccccc24)cc1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H22ClN3O/c22-16-9-7-15(8-10-16)14-20-18-5-1-2-6-19(18)21(26)25(24-20)17-4-3-12-23-13-11-17/h1-2,5-10,17,23H,3-4,11-14H2
Formula Weight 367.1451
Chemical Name or Material N-Desmethyl Azelastine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.