missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl imatinib, MedChemExpress

Product Code. 16481228
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481228 5 mg 5mg
16478798 10 mg 10mg
16464707 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481228 Supplier Medchem Express Supplier No. HYG0017
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress N-Desmethyl imatinib (Norimatinib) is a metabolite of Imatinib (HY-15463). Imatinib is a multi-target inhibitor of v-Abl, c-Kit and PDGFR.

N-Desmethyl imatinib (Norimatinib) is a metabolite of Imatinib (HY-15463). Imatinib is a multi-target inhibitor of v-Abl, c-Kit and PDGFR.

Specifications

Chemical Name or Material N-Desmethyl imatinib
CAS 404844-02-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H29N7O
Quantity 5 mg
Synonym Norimatinib Imatinib metabolite N-Desmethyl imatinib
Solubility Information DMSO : 100 mg/mL (208.52 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C)C(NC2=NC(C3=CC=CN=C3)=CC=N2)=C1)C4=CC=C(CN5CCNCC5)C=C4
Molecular Weight (g/mol) 479.58
Formula Weight 479.58
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.