missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl N-Nitroso Imatinib, TRC

Product Code. 30477993
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477993 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477993 Supplier Toronto Research Chemicals Supplier No. TRCM45878510MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Desmethyl N-Nitroso Imatinib
CAS 2816369-80-7
Molecular Formula C28 H28 N8 O2
Quantity 10 mg
Synonym N-(4-Methyl-3((4-(pyridin-3yl)pyrimidin-2-yl)amino)phenyl)-4-((4-nitrosopiperazin-1-yl)methyl)benzamide
InChI Formula InChI=1S/C28H28N8O2/c1-20-4-9-24(17-26(20)33-28-30-12-10-25(32-28)23-3-2-11-29-18-23)31-27(37)22-7-5-21(6-8-22)19-35-13-15-36(34-38)16-14-35/h2-12,17-18H,13-16,19H2,1H3,(H,31,37)(H,30,32,33)
SMILES Cc1ccc(NC(=O)c2ccc(CN3CCN(CC3)N=O)cc2)cc1Nc4nccc(n4)c5cccnc5
IUPAC Name N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]-4-[(4-nitrosopiperazin-1-yl)methyl]benzamide
Molecular Weight (g/mol) 508.574
Formula Weight 508.234
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.