missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethylclozapine, MedChemExpress

Product Code. 16498798
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498798 10 mg 10mg
16401238 5 mg 5mg
16433826 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498798 Supplier Medchem Express Supplier No. HYG0021
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress N-Desmethylclozapine is a major active metabolite of the atypical antipsychotic drug Clozapine. N-Desmethylclozapine is a potent, allosteric and partial M1 receptors agonist (EC50=115 nM) and is able to potentiate hippocampal N-methyl-d-aspartate (NMDA) receptor currents through M1 receptor activation. N-Desmethylclozapine is also a δ-opioid agonist.

N-Desmethylclozapine is a major active metabolite of the atypical antipsychotic drug Clozapine. N-Desmethylclozapine is a potent, allosteric and partial M1 receptors agonist (EC50=115 nM) and is able to potentiate hippocampal N-methyl-d-aspartate (NMDA) receptor currents through M1 receptor activation. N-Desmethylclozapine is also a δ-opioid agonist.

Specifications

Chemical Name or Material N-Desmethylclozapine
CAS 6104-71-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H17ClN4
Quantity 10 mg
Synonym Norclozapine Desmethylclozapine Normethylclozapine
Solubility Information DMSO : ≥ 50 mg/mL (159.85 mM)
SMILES ClC1=CC=C2C(N=C(N3CCNCC3)C4=CC=CC=C4N2)=C1
Molecular Weight (g/mol) 312.8
Formula Weight 312.8
Percent Purity 99.66%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.