missing translation for 'onlineSavingsMsg'
Learn More

n-Docosane, 99%

Product Code. 11409196
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11409196 25 g 25g
11419196 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11409196 Supplier Thermo Scientific Alfa Aesar Supplier No. A18050.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 629-97-0 | C22H46 | 310.61 g/mol

It is employed as a perfuming agent in cosmetics industry. It is also used in organic synthesis, calibration, and temperature sensing equipment. Docosane is used to investigate commercially available waxes in the form of thin disc samples as possible diffraction intensity standards for macromolecular crystallography synchrotron beamlines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a perfuming agent in cosmetics industry. It is also used in organic synthesis, calibration, and temperature sensing equipment. Docosane is used to investigate commercially available waxes in the form of thin disc samples as possible diffraction intensity standards for macromolecular crystallography synchrotron beamlines.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-97-0
Molecular Formula C22H46
Molecular Weight (g/mol) 310.61
MDL Number MFCD00009348
InChI Key HOWGUJZVBDQJKV-UHFFFAOYSA-N
Synonym n-docosane, dokosan, unii-ow99q363ko, heneicosane, methyl, ch3-ch2 20-ch3, docosane, analytical standard, twt, normal-docosane, docosane, n, docosane
PubChem CID 12405
ChEBI CHEBI:46050
IUPAC Name docosane
SMILES CCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 43°C to 46°C
Density 0.777
Boiling Point 368°C to 369°C
Flash Point >110°C (230°F)
Refractive Index 1.4455
Quantity 25 g
Beilstein 1702206
Solubility Information Insoluble in water. Soluble in alcohol.
Formula Weight 310.61
Percent Purity 99%
Chemical Name or Material n-Docosane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.