missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecane, 99%, anhydrous, AcroSeal™

Product Code. 11924411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11924411 100 mL AcroSeal™ Glass Bottle 100mL
11914411 1 L AcroSeal™ Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11924411 Supplier Thermo Scientific Acros Supplier No. 434591000
  • ONLINE EXCLUSIVE PRICE
    111.00 €
    101.00 € / 100mL
    Save 10.00 €
    9% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 112-40-3 | C12H26 | 170.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Melting Point -12.0°C
Color Colorless
Density 0.7530g/mL
Boiling Point 215.0°C to 217.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3(CH2)10CH3
Refractive Index 1.4210 to 1.4230
Quantity 100 mL
Specific Gravity 0.753
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether
Viscosity <7 mm2/s (40°C)
Formula Weight 170.34
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Dodecane
Show More Show Less
compliance-icons
Product Identifier
  • n-Dodecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: Dodecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.