Product Code. 10318393
Shop All Fisher Chemical Products
Click to view available options
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
10318393
2.5 L
2.5 L
10142090
100 mL
100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
10318393
Supplier
Fisher Chemical
Supplier No.
D/5290/17
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Chemical Identifiers
CAS
112-40-3
Molecular Formula
C12H26
Molecular Weight (g/mol)
170.34
MDL Number
8969
InChI Key
SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym
n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID
8182
ChEBI
CHEBI:28817
IUPAC Name
dodecane
SMILES
CCCCCCCCCCCC
Specifications
Melting Point
-12°C
Color
Colorless
Boiling Point
215°C
Packaging
Amber glass bottle
Quantity
2.5 L
Vapor Density
&5.9
Formula Weight
170.34g/mol
Physical Form
Liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?