missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl acrylate, tech. 90%, stab. with 60-100ppm 4-methoxyphenol

Product Code. 15495896
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495896 500 mL 500mL
15485896 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495896 Supplier Thermo Scientific Alfa Aesar Supplier No. H60625.AP
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2156-97-0 | C15H28O2 | 240.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2156-97-0
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.39
MDL Number MFCD00042871
InChI Key PBOSTUDLECTMNL-UHFFFAOYSA-N
Synonym dodecyl acrylate, lauryl acrylate, n-lauryl acrylate, n-dodecyl acrylate, 2-propenoic acid, dodecyl ester, acrylic acid, dodecyl ester, acrylic acid lauryl ester, acrylic acid dodecyl ester, unii-90ik36wgvr, 90ik36wgvr
PubChem CID 75084
IUPAC Name dodecyl prop-2-enoate
SMILES CCCCCCCCCCCCOC(=O)C=C

Spécification

Density 0.884
Boiling Point 120°C (0.75 mmHg)
Flash Point 110°C (230°F)
Linear Formula CH2=CHCOOC12H25
Refractive Index 1.445
Quantity 500 mL
UN Number UN3082
Beilstein 1779069
Formula Weight 240.38
Percent Purity 90%
Grade Technical
Chemical Name or Material n-Dodecyl acrylate, stabilized with 60 to 100ppm 4-methoxyphenol
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.