missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl beta-D-Maltoside, TRC

Product Code. 30137747
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137747 1 g 1g
30136290 5 g 5g
30006324 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137747 Supplier Toronto Research Chemicals Supplier No. TRCD5253551G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69227-93-6
Molecular Formula C24 H46 O11
Molecular Weight (g/mol) 510.62
Synonym n-Dodecyl β-D-Maltoside, 1-Dodecyl-β-D-maltoside, DDM, DDM (Detergent), Dodecyl O-β-D-Maltoside, Dodecyl β-D-Maltopyranoside, Dodecyl β-D-Maltoside, Dodecyl β-Maltopyranoside, Dodecyl β-Maltoside, Lauryl Maltoside, Lauryl β-D-Maltopyranoside, Lauryl β-D-Maltoside, Lauryl β-Maltopyranoside, Lauryl β-Maltoside, n-Dodecyl β-D-Maltopyranoside, β-1-n-Dodecyl-D-Maltoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6S)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6S)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)tetrahydropyran-3-yl]oxy-6-(hydroxymethyl)tetrahydropyran-3,4,5-triol
SMILES CCCCCCCCCCCCO[C@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C24H46O11/c1-2-3-4-5-6-7-8-9-10-11-12-32-23-21(31)19(29)22(16(14-26)34-23)35-24-20(30)18(28)17(27)15(13-25)33-24/h15-31H,2-14H2,1H3/t15-,16-,17-,18+,19-,20-,21-,22-,23+,24-/m1/s1
Formula Weight 510.304
Percent Purity >95%
Chemical Name or Material n-Dodecyl beta-D-Maltoside
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.