missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl-beta-D-maltoside, 99%, high purity

Product Code. 10207353 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10207353 5 g Glass bottle 5g
10376221 1 g Glass bottle 1g
10415555 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10207353 Supplier Thermo Scientific Acros Supplier No. 329370050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 69227-93-6 | C24H46O11 | 510.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69227-93-6
Molecular Formula C24H46O11
Molecular Weight (g/mol) 510.6
MDL Number MFCD00043012
InChI Key NLEBIOOXCVAHBD-QKMCSOCLSA-N
Synonym n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside
PubChem CID 114880
ChEBI CHEBI:43769
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 217.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, V,8, 12
Specific Rotation Condition +47.50 (20.00°C c=1,water)
Specific Rotation + 47.50
Solubility Information Solubility in water: soluble (100g/L). Other solubilities: soluble in methanol, ethanol
Formula Weight 510.6
Percent Purity 99%
Grade High Purity
Physical Form Powder
Chemical Name or Material n-Dodecyl-β-D-maltoside
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.