missing translation for 'onlineSavingsMsg'
Learn More

N-Dodecyl-β-D-Maltoside, Approx. 97%, MP Biomedicals™

Product Code. 17841371 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17841371 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17841371 Supplier MP Biomedicals Supplier No. 0215101380
  • ONLINE EXCLUSIVE PRICE
    47.65 €
    45.15 € / 100mg
    Save 2.50 €
    5% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color White
CAS 69227-93-6
Physical Form Solid
Quantity 100 mg
Assay Percent Range Approx. 97%
Molecular Formula C24H46O11
MDL Number MFCD00043012
Synonym n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside
InChI Key NLEBIOOXCVAHBD-QKMCSOCLSA-N
SMILES CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 510.621
PubChem CID 114880
ChEBI CHEBI:43769
Formula Weight 510.6
Chemical Name or Material N-Dodecyl-β-D-Maltoside
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.