missing translation for 'onlineSavingsMsg'
Learn More

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate

Product Code. 15490537
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490537 25 g 25g
15480537 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490537 Supplier Thermo Scientific Alfa Aesar Supplier No. J66265.14
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

For protein solubilization | CAS: 14933-08-5 | C17H37NO3S | 335.547 g/mol

For protein solubilization

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14933-08-5
Molecular Formula C17H37NO3S
Molecular Weight (g/mol) 335.547
MDL Number MFCD00036909
InChI Key IZWSFJTYBVKZNK-UHFFFAOYSA-N
Synonym lauryl sultaine, lauryl sulfobetaine, n-dodecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dodecyldimethylammonio propane-1-sulfonate, zwittergent 3-12, 3-dodecyldimethylammonio propanesulfonate, sulfobetaine 12, unii-r6p3kw3e8u, 3-lauryldimethylammonio propanesulfonate, dodecyldimethyl 3-sulphonatopropyl ammonium
PubChem CID 84703
ChEBI CHEBI:75303
IUPAC Name 3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

Color White
Flash Point 110°C (230°F)
Quantity 25 g
Beilstein 4145308
Sensitivity Hygroscopic
Solubility Information Soluble at 1M in water.
Formula Weight 335.55
Physical Form Powder
Chemical Name or Material N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.