missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ n-Dodecyl thiocyanate, 97%

Product Code. 11311757
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11311757

Brand: Alfa Aesar™ L04902.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 765-15-1
Molecular Formula C13H25NS
Molecular Weight (g/mol) 227.41
MDL Number MFCD00019773
InChI Key RYZZMYXTNUJGMU-UHFFFAOYSA-N
Synonym lauryl thiocyanate, n-dodecyl thiocyanate, thiocyanic acid, dodecyl ester, lauryl rhodanate, lorol thiocyanate, laurylrhodanide, loro, dodecane, 1-thiocyanato, laurylthiokyanat, 1-dodecyl thiocyanate
PubChem CID 69827
IUPAC Name dodecyl thiocyanate
SMILES CCCCCCCCCCCCSC#N

Specifications

Density 0.896
Boiling Point 180°C (18 mmHg)
Refractive Index 1.468
Quantity 5 g
Beilstein 1770396
Formula Weight 227.41
Percent Purity 97%
Chemical Name or Material n-Dodecyl thiocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.