missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ n-Dodecyl thiocyanate, 97%

Product Code. 11331757
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331757 100 g 100g
11311757 5 g 5g
11321757 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331757 Supplier Alfa Aesar™ Supplier No. L04902.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 765-15-1
Molecular Formula C13H25NS
Molecular Weight (g/mol) 227.41
MDL Number MFCD00019773
InChI Key RYZZMYXTNUJGMU-UHFFFAOYSA-N
Synonym lauryl thiocyanate, n-dodecyl thiocyanate, thiocyanic acid, dodecyl ester, lauryl rhodanate, lorol thiocyanate, laurylrhodanide, loro, dodecane, 1-thiocyanato, laurylthiokyanat, 1-dodecyl thiocyanate
PubChem CID 69827
IUPAC Name dodecyl thiocyanate
SMILES CCCCCCCCCCCCSC#N

Specifications

Density 0.896
Boiling Point 180°C (18 mmHg)
Refractive Index 1.468
Quantity 100 g
Beilstein 1770396
Formula Weight 227.41
Percent Purity 97%
Chemical Name or Material n-Dodecyl thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.