missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecylbenzene, 97%

Product Code. p-7025720
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11499297

Brand: Thermo Scientific Alfa Aesar B21362.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-01-3 | C18H30 | 246.438 g/mol

n-Dodecylbenzene, is a precursor to sodium dodecylbenzenesulfonate, a surfactant that is a key ingredient of household laundry detergents, such as Tide. It is a widely used cleaner, dyeing assistant in textile industrial, degrease agent in electroplate, leather, de- inking agent in papering, rubber and plastic, break emulsion agent in oil industry, artesian well foaming agent in oil well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Dodecylbenzene, is a precursor to sodium dodecylbenzenesulfonate, a surfactant that is a key ingredient of household laundry detergents, such as Tide. It is a widely used cleaner, dyeing assistant in textile industrial, degrease agent in electroplate, leather, de- inking agent in papering, rubber and plastic, break emulsion agent in oil industry, artesian well foaming agent in oil well.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, nitric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-01-3
Molecular Formula C18H30
Molecular Weight (g/mol) 246.438
MDL Number MFCD00008974
InChI Key KWKXNDCHNDYVRT-UHFFFAOYSA-N
Synonym 1-phenyldodecane, benzene, dodecyl, n-dodecylbenzene, laurylbenzene, phenyldodecane, marlican, dodecane, 1-phenyl, detergent alkylate, alkylate p 1, phenyldodecan
PubChem CID 31237
IUPAC Name dodecylbenzene
SMILES CCCCCCCCCCCCC1=CC=CC=C1

Specifications

Melting Point -7°C
Density 0.856
Boiling Point 331°C
Flash Point 124°C (255°F)
Refractive Index 1.482
Quantity 5 g
Beilstein 1909107
Solubility Information Insoluble in water.
Formula Weight 246.44
Percent Purity 97%
Chemical Name or Material n-Dodecylbenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.