missing translation for 'onlineSavingsMsg'
Learn More

N-(Ethoxycarbonyl)nortropinone, 99%, Thermo Scientific™

Product Code. 11303287
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303287 5 g 5g
11393277 1 g 1g
11313287 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11303287 Supplier Thermo Scientific Alfa Aesar Supplier No. L13849.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32499-64-2
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.234
MDL Number MFCD00078171
InChI Key ANEJUHJDPGTVIO-UHFFFAOYSA-N
Synonym ethyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-ethoxycarbonyl nortropinone, n-carbethoxy-4-nortropinone, n-carbethoxy-4-tropinone, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one, 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid ethyl ester, 8-azabicyclo 3.2.1 octane-8-carboxylic acid, 3-oxo-, ethyl ester, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, n-carbethoxynortropinone
PubChem CID 401882
IUPAC Name ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
SMILES CCOC(=O)N1C2CCC1CC(=O)C2

Specifications

Boiling Point 152°C to 153°C (11 mmHg)
Refractive Index 1.4895
Quantity 5 g
Beilstein 1530944
Formula Weight 197.23
Percent Purity 99%
Chemical Name or Material N-(Ethoxycarbonyl)nortropinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.