missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl Fluprostenol Carboxamide, TRC

Product Code. 30381174
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381174 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381174 Supplier Toronto Research Chemicals Supplier No. TRCE9181505MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Ethyl Fluprostenol Carboxamide
CAS 1005193-64-5
Molecular Formula C25H34F3NO5
Quantity 5 mg
Synonym (5Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(1E,3R)-3-hydroxy-4-[3-(trifluoromethyl)phenoxy]-1-buten-1-yl]cyclopentyl]-N-ethyl-5-heptenamide
InChI Formula InChI=1S/C25H34F3NO5/c1-2-29-24(33)11-6-4-3-5-10-20-21(23(32)15-22(20)31)13-12-18(30)16-34-19-9-7-8-17(14-19)25(26,27)28/h3,5,7-9,12-14,18,20-23,30-32H,2,4,6,10-11,15-16H2,1H3,(H,29,33)/b5-3-,13-12+/t18-,20+,21+,22-,23+/m0/s1
SMILES O[C@@H]1[C@H](C/C=C\CCCC(NCC)=O)[C@@H](/C=C/[C@@H](O)COC2=CC=CC(C(F)(F)F)=C2)[C@H](O)C1
IUPAC Name (Z)-7-((1R,2R,3R,5S)-3,5-dihydroxy-2-((R,E)-3-hydroxy-4-(3-(trifluoromethyl)phenoxy)but-1-en-1-yl)cyclopentyl)-N-ethylhept-5-enamide
Molecular Weight (g/mol) 485.54
Formula Weight 485.24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.