missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-hydroxyethylaniline, 96%

Product Code. 10442271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10442271 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10442271 Supplier Thermo Scientific Acros Supplier No. 325221000
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-50-2 | C10H15NO | 165.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 92-50-2
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Orange
Density 1.02g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C10H15NO
Linear Formula C6H5N(CH5)CHCHOH
Refractive Index 1.5600 to 1.5640 (20°C, 589nm)
MDL Number MFCD00020575
Quantity 100 g
Synonym 2-n-ethylanilino ethanol, 2-ethyl phenyl amino ethanol, ethanol, 2-ethylphenylamino, ethylphenylethanolamine, phenylethylethanolamine, n-ethyl-n-2-hydroxyethyl aniline, n-ethylanilinoethanol, hydroxyethylethylaniline, n-ethyl-n-hydroxyethylaniline, 2-ethylphenylamino ethanol
InChI Key HYVGFUIWHXLVNV-UHFFFAOYSA-N
SMILES CCN(CCO)C1=CC=CC=C1
IUPAC Name 2-[ethyl(phenyl)amino]ethan-1-ol
Molecular Weight (g/mol) 165.24
PubChem CID 62338
Formula Weight 165.24
Percent Purity ≥95% (GC)
Physical Form Liquid
Chemical Name or Material N-Ethyl-N-hydroxyethylaniline, 96%
Show More Show Less
compliance-icons
Product Identifier
  • N-Ethyl-N-hydroxyethylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.