missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-nitroso-2-propanamine, TRC

Product Code. 17993945
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17993945 25 mg 25mg
17922365 100 mg 100mg
17951485 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993945 Supplier Toronto Research Chemicals Supplier No. TRCE93279525MG
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16339-04-1
Molecular Formula C5 H12 N2 O
Molecular Weight (g/mol) 116.1616
Synonym 2-Propanamine, N-ethyl-N-nitroso-, Diethylamine, 1-methyl-N-nitroso- (7CI,8CI), N-Ethyl-N-nitroso-2-propanamine, 1-Methyl-N-nitrosodiethylamine, Ethylisopropylnitrosamine, EIPNA, N-Nitroso-N-ethyl-N-isopropylamine, NEIPA, NEPA
IUPAC Name N-ethyl-N-propan-2-ylnitrous amide
SMILES CCN(N=O)C(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H12N2O/c1-4-7(6-8)5(2)3/h5H,4H2,1-3H3
Formula Weight 116.095
Percent Purity >95%
Chemical Name or Material N-Ethyl-N-nitroso-2-propanamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.