missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylcyclohexylamine, 97%

Product Code. 11319055
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319055 100 mL 100mL
11309055 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319055 Supplier Thermo Scientific Alfa Aesar Supplier No. L14221.AE
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5459-93-8 | C8H17N | 127.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5459-93-8
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD00003834
InChI Key AGVKXDPPPSLISR-UHFFFAOYSA-N
Synonym n-ethylcyclohexylamine, cyclohexanamine, n-ethyl, n-cyclohexylethylamine, vulkacit hx, accelerator hx, cyclohexylamine, n-ethyl, ethylamino cyclohexane, cyclohexyl ethyl amine, n-ethyl cyclohexyl amine, unii-yjk13p0h3e
PubChem CID 21609
IUPAC Name N-ethylcyclohexanamine
SMILES CCNC1CCCCC1

Specifications

Density 0.85
Boiling Point 163°C to 166°C
Flash Point 45°C (113°F)
Odor Amine-like
Refractive Index 1.452
Quantity 100 mL
UN Number UN2734
Beilstein 2070280
Sensitivity Air Sensitive
Formula Weight 127.23
Percent Purity 97%
Chemical Name or Material N-Ethylcyclohexylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.