missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylcyclohexylamine, 98%, Thermo Scientific™

Product Code. 11386365 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11386365 25 mL Glass bottle 25mL
11396365 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386365 Supplier Thermo Scientific Acros Supplier No. 437740250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5459-93-8
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
InChI Key AGVKXDPPPSLISR-UHFFFAOYSA-N
Synonym n-ethylcyclohexylamine, cyclohexanamine, n-ethyl, n-cyclohexylethylamine, vulkacit hx, accelerator hx, cyclohexylamine, n-ethyl, ethylamino cyclohexane, cyclohexyl ethyl amine, n-ethyl cyclohexyl amine, unii-yjk13p0h3e
PubChem CID 21609
IUPAC Name N-ethylcyclohexanamine
SMILES CCNC1CCCCC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8500g/mL
Boiling Point 164°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4500 to 1.4530 (20°C, 589nm)
Quantity 25 mL
Specific Gravity 0.85
Formula Weight 127.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Ethylcyclohexylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Ethylcyclohexylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.