missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmaleimide, 98+%

Product Code. 11340014
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11340014 50 g 50g
11330014 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340014 Supplier Thermo Scientific Alfa Aesar Supplier No. L00355.18
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 128-53-0 | C6H7NO2 | 125.127 g/mol

Cysteine alkylating agent N-Ethylmaleimide is a protein thiol modifier which inhibits apoptotic DNA fragmentation. It is a sulfhydryl reagent and finds application in experimental biochemical studies as well as in enzymology. It is involved in the modification of cysteine residues in proteins and peptides. It acts as a Michael acceptor and reacts with nucelophiles like thiols. It is employed in the inhibition of Mg2+-dependent internucleosomal DNA fragmentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cysteine alkylating agent N-Ethylmaleimide is a protein thiol modifier which inhibits apoptotic DNA fragmentation. It is a sulfhydryl reagent and finds application in experimental biochemical studies as well as in enzymology. It is involved in the modification of cysteine residues in proteins and peptides. It acts as a Michael acceptor and reacts with nucelophiles like thiols. It is employed in the inhibition of Mg2+-dependent internucleosomal DNA fragmentation.

Solubility
Soluble in ethanol, water, chloroform, methanol and toluene.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-53-0
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00005509
InChI Key HDFGOPSGAURCEO-UHFFFAOYSA-N
Synonym n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
PubChem CID 4362
ChEBI CHEBI:44485
IUPAC Name 1-ethylpyrrole-2,5-dione
SMILES CCN1C(=O)C=CC1=O

Specifications

Melting Point 43°C to 46°C
Boiling Point 209°C to 211°C
Flash Point 73°C (163°F)
Odor Pungent
Quantity 50 g
UN Number UN2928
Beilstein 112448
Sensitivity Light sensitive
Merck Index 14,3822
Solubility Information Soluble in ethanol,water,chloroform,methanol and toluene.
Formula Weight 125.13
Percent Purity ≥98%
Chemical Name or Material N-Ethylmaleimide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.