missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmaleimide, MedChemExpress

Product Code. 16458768
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458768 Supplier Medchem Express Supplier No. HYD0843
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress N-Ethylmaleimide (NEM) derives from maleic acid, it can alkylates free sulfhydryl. N-Ethylmaleimide is an irreversible cysteine protease inhibitor. N-ethylmaleimide specific inhibits phosphate transport in mitochondria. N-Ethylmaleimide inhibits prolyl endopeptidase with an IC50 value of 6.3 μM. N-Ethylmaleimide can be used to modify cysteine residues in proteins and peptides.

N-Ethylmaleimide (NEM) derives from maleic acid, it can alkylates free sulfhydryl. N-Ethylmaleimide is an irreversible cysteine protease inhibitor. N-ethylmaleimide specific inhibits phosphate transport in mitochondria. N-Ethylmaleimide inhibits prolyl endopeptidase with an IC50 value of 6.3 μM. N-Ethylmaleimide can be used to modify cysteine residues in proteins and peptides.

Specifications

Chemical Name or Material N-Ethylmaleimide
CAS 128-53-0
Color White
Purity Grade Notes Research
Molecular Formula C6H7NO2
Quantity 10 mg
Synonym NEM
Solubility Information H2O : 50 mg/mL (399.58 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (399.58 mM; Need ultrasonic) ∣Ethanol : 12.5 mg/mL (99.90 mM; Need ultrasonic)
SMILES O=C(C=C1)N(CC)C1=O
Molecular Weight (g/mol) 125.13
Formula Weight 125.13
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.