missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmethylamine, TRC

Product Code. 30135789
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135789 5 g 5g
30136985 50 g 50g
30010098 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135789 Supplier Toronto Research Chemicals Supplier No. TRCE9248505G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-78-2
Molecular Formula C3 H9 N
Molecular Weight (g/mol) 59.11
Synonym Ethanamine, N-methyl- (9CI, ACI), Ethylamine, N-methyl- (7CI, 8CI), N-Methylethanamine (ACI), Ethylmethylamine, Methylaminoethane, Methylethylamine, N-Ethyl-N-methylamine, N-Ethylmethylamine, N-Methyl-N-ethylamine, N-Methylethylamine, NMEA
IUPAC Name N-methylethanamine
SMILES CCNC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H9N/c1-3-4-2/h4H,3H2,1-2H3
Formula Weight 59.0735
Chemical Name or Material N-Ethylmethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.