missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmorpholine, 99%

Product Code. 10274810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10274810 100 mL Glass bottle 100mL
10745041 500 mL Glass bottle 500mL
10227213 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10274810 Supplier Thermo Scientific Acros Supplier No. 149401000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-74-3 | C6H13NO | 115.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 100-74-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
MDL Number MFCD00006177
InChI Key HVCNXQOWACZAFN-UHFFFAOYSA-N
Synonym n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq
PubChem CID 7525
IUPAC Name 4-ethylmorpholine
SMILES CCN1CCOCC1

Spécification

CAS Min % 98.5
CAS Max % 100.0
Melting Point -63.0°C
Color Brown-Gray to Cream
Density 0.9000g/mL
Boiling Point 139.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420
Quantity 100 mL
Beilstein 27, 6
Fieser 01,383
Specific Gravity 0.9
Solubility Information Solubility in water: miscible with water. Other solubilities: soluble in alcohol,ether,acetone and benzene
Formula Weight 115.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-Ethylmorpholine, 99%
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • N-Ethylmorpholine
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Mention de danger
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contain: N-Ethylmorpholine

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.