missing translation for 'onlineSavingsMsg'
Learn More

N-Fluoropyridinium triflate, 95%, ACROS Organics™

Product Code. 10083340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10083340 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10083340 Supplier Thermo Scientific Acros Supplier No. 259925000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 107263-95-6
CAS Min % 94.0
CAS Max % 100.0
Melting Point 182°C to 187°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C6H5F4NO3S
MDL Number MFCD00013458
Quantity 500 mg
Fieser 15,160; 16,170
Synonym 1-fluoropyridinium triflate, n-fluoropyridinium triflate, n-fluoropyridinium trifluoromethanesulfonate, 1-fluoropyridinium trifluoromethanesulfonate, 1-fluoropyridin-1-ium; trifluoromethanesulfonate, acmc-209shu, ksc182s3j, 1-fluoropyridin-1-ium triflate, 1-fluoropyridiniumtriflate
InChI Key JFZMMCYRTJBQQI-UHFFFAOYSA-M
SMILES C1=CC=[N+](C=C1)F.C(F)(F)(F)S(=O)(=O)[O-]
IUPAC Name 1-fluoropyridin-1-ium;trifluoromethanesulfonate
Molecular Weight (g/mol) 247.164
PubChem CID 2724576
Formula Weight 247.16
Percent Purity ≥94%
Physical Form Crystalline Powder or Needles
Chemical Name or Material N-Fluoropyridinium triflate, 95%
Show More Show Less
compliance-icons
Product Identifier
  • N-Fluoropyridinium triflate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.