missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-chloro-L-phenylalanine, 98%

Product Code. 15420149
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420149 5 g 5g
15430149 250 mg 250mg
15410149 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420149 Supplier Thermo Scientific Alfa Aesar Supplier No. H63448.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 175453-08-4 | C24H20ClNO4 | 421.877 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175453-08-4
Molecular Formula C24H20ClNO4
Molecular Weight (g/mol) 421.877
MDL Number MFCD00080281
InChI Key CQPNKLNINBUUOM-QFIPXVFZSA-N
Synonym fmoc-4-chloro-l-phenylalanine, s-n-fmoc-4-chlorophenylalanine, fmoc-phe 4-cl-oh, fmoc-l-4-chlorophenylalanine, fmoc-l-phe 4-cl-oh, fmoc-4-chloro-phe-oh, fmoc-l-4-chlorophe, 2s-3-4-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid, phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl
PubChem CID 2734471
IUPAC Name (2S)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O

Specifications

Beilstein 9437838
Quantity 5 g
Formula Weight 421.87
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-4-chloro-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.