missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-alanine, TRC

Product Code. 30179551
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179551 5 g 5g
30178999 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179551 Supplier Toronto Research Chemicals Supplier No. TRCF6199555G
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79990-15-1
Molecular Formula C18 H17 N O4
Molecular Weight (g/mol) 311.33
Synonym N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine, (R)-2-[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]propionic Acid, (R)-N-Fmoc-alanine, 270: PN: US20070042401 PAGE: 36 Claimed Protein, 962: PN: WO2006135786 PAGE: 69 Claimed Protein, D-Fmoc-Alanine, N-(9-Fluorenylmethoxycarbonyl)-D-alanine
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C[C@@H](NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O

Specifications

InChI Formula InChI=1S/C18H17NO4/c1-11(17(20)21)19-18(22)23-10-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,11,16H,10H2,1H3,(H,19,22)(H,20,21)/t11-/m1/s1
Quantity 5 g
Formula Weight 311.116
Shipping Condition Room Temperature
Chemical Name or Material N-Fmoc-D-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.