missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-allo-isoleucine, 98%

Product Code. 15460159
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460159 5 g 5g
15470159 250 mg 250mg
15450159 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15460159 Supplier Thermo Scientific Alfa Aesar Supplier No. H63455.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118904-37-3 | C21H23NO4 | 353.42 g/mol

N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate at -20°C. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118904-37-3
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00077062
InChI Key QXVFEIPAZSXRGM-KINVLMLONA-N
Synonym fmoc-d-allo-ile-oh, fmoc-d-allo-isoleucine, fmoc-d-allo-lle-oh, 2r,3s-n-fmoc-2-amino-3-methylpentanoic acid, 2r,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, d-alloisoleucine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r,3s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylpentanoic acid, ambotzfaa1328, pubchem15619
PubChem CID 6992517
IUPAC Name (2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylpentanoic acid
SMILES CC[C@H](C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 353.41
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-D-allo-isoleucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.