missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-methionine, 98%, Thermo Scientific™

Product Code. 15448738
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448738 100 g 100g
15428738 5 g 5g
15438738 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448738 Supplier Thermo Scientific Alfa Aesar Supplier No. H62178.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112883-40-6
Molecular Formula C20H21NO4S
Molecular Weight (g/mol) 371.45
MDL Number MFCD00062958 MFCD00037134 MFCD00062958
InChI Key BUBGAUHBELNDEW-GOSISDBHSA-N
Synonym fmoc-d-met-oh, fmoc-d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl-d-methionine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylthio butanoic acid, d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, n-fmoc-d-methionine, ambotzfaa1145, n-9-fluorenylmethyloxycarbonyl-d-methionine
PubChem CID 6992521
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfanylbutanoic acid
SMILES CSCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 130°C to 134°C
Quantity 100 g
Formula Weight 371.45
Optical Rotation +30° (c=1 in DMF)
Percent Purity 98%
Chemical Name or Material N-Fmoc-D-methionine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.