missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-methionine, TRC

Product Code. 30406528
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406528 25 g 25g
30389483 100 g -
30399425 250 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406528 Supplier Toronto Research Chemicals Supplier No. TRCF62285025G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112883-40-6
Molecular Formula C20H21NO4S
Molecular Weight (g/mol) 371.45
Synonym N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine
IUPAC Name (((9H-fluoren-9-yl)methoxy)carbonyl)-D-methionine
SMILES O=C(O)[C@H](NC(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)=O)CCSC

Specifications

InChI Formula InChI=1S/C20H21NO4S/c1-26-11-10-18(19(22)23)21-20(24)25-12-17-15-8-4-2-6-13(15)14-7-3-5-9-16(14)17/h2-9,17-18H,10-12H2,1H3,(H,21,24)(H,22,23)/t18-/m1/s1
Quantity 25 g
Formula Weight 371.12
Shipping Condition Room Temperature
Chemical Name or Material N-Fmoc-D-methionine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.