missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-L-valine, 95%

Product Code. 15480479
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480479 1 g 1g
15490479 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480479 Supplier Thermo Scientific Alfa Aesar Supplier No. H63671.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 84000-11-3 | C21H23NO4 | 353.42 g/mol

N-Fmoc-N-methyl-L-valine is used as enzyme substrates and reagents, culture media additives, dyes, stains and indicators. It is also involved in the preparation of N-methylated peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-N-methyl-L-valine is used as enzyme substrates and reagents, culture media additives, dyes, stains and indicators. It is also involved in the preparation of N-methylated peptides.

Solubility
Slightly soluble in water and dimethyl formamide.

Notes
Incompatible with strong oxidizing agents, reducing agents, bases and alkali metals. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84000-11-3
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00153395
InChI Key YCXXXPZNQXXRIG-IBGZPJMESA-N
Synonym fmoc-n-me-val-oh, fmoc-n-methyl-l-valine, n-fmoc-n-methyl-l-valine, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-methylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl-l-valine, l-valine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-3-methyl-butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylbutanoic acid, ambotzfaa1411, fmoc-n-me-l-val-oh
PubChem CID 978328
IUPAC Name (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylbutanoic acid
SMILES CC(C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 187°C to 190°C
Color White
Solubility Information Slightly soluble in water and dimethyl formamide.
Quantity 1 g
Formula Weight 353.41
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-N-methyl-L-valine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.