missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-tert-butyl-L-tyrosine, 98%

Product Code. 15476128
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15476128

Brand: Thermo Scientific Alfa Aesar H59730.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71989-38-3 | C28H29NO5 | 459.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-38-3
Molecular Formula C28H29NO5
Molecular Weight (g/mol) 459.54
MDL Number MFCD00037129
InChI Key JAUKCFULLJFBFN-KSYWNVGFNA-N
Synonym fmoc-tyr tbu-oh, fmoc-o-tert-butyl-l-tyrosine, fmoc-tyr but-oh, fmoc-tyr t-bu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, fmoc-tyr but, n-fmoc-o-tert-butyl-l-tyrosine, nalpha-fmoc-o-tert-butyl-l-tyrosine, n-9-fluorenylmethoxy carbonyl-o-tert-butyl-l-tyrosine, 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
PubChem CID 10895791
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid
SMILES CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1

Specifications

Melting Point 153°C to 156°C
Quantity 25 g
Beilstein 4216652
Optical Rotation −29° (c=1 in DMF)
Formula Weight 459.53
Percent Purity 98%
Chemical Name or Material N-Fmoc-O-tert-butyl-L-tyrosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.