missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-methionine, 95%

Product Code. 15400219
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
missing translation for 'unitSize':
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
15400219 1 g 1g
15420219 250 mg 250mg
15410219 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15400219 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' H63493.03
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4289-98-9 | C6H11NO3S | 177.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4289-98-9
Molecular Formula C6H11NO3S
Molecular Weight (g/mol) 177.22
MDL Number MFCD00021033
InChI Key PYUSHNKNPOHWEZ-YFKPBYRVSA-N
Synonym n-formyl-l-methionine, n-formylmethionine, for-met-oh, s-2-formamido-4-methylthio butanoic acid, formyl-l-methionine, formylmethionine, l-methionine, n-formyl, unii-ps9357b4xh, n-formyl methionine
PubChem CID 439750
ChEBI CHEBI:16552
IUPAC Name (2S)-2-formamido-4-methylsulfanylbutanoic acid
SMILES CSCC[C@H](NC=O)C(O)=O

Specifications

Quantity 1 g
Formula Weight 177.22
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Formyl-L-methionine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.