missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-proline, 97%

Product Code. 15462162 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462162 1 g 1g
15472162 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462162 Supplier Thermo Scientific Acros Supplier No. 456420010
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13200-83-4 | C6H9NO3 | 143.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 13200-83-4
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.14
InChI Key DHDRGOURKDLAOT-YFKPBYRVSA-N
Synonym 1-formyl-l-proline, l-proline, 1-formyl, n-formylproline, n-formyl-l-proline, 2s-1-formylpyrrolidine-2-carboxylic acid, s---n-formylproline, n-formyl-l-proline tlc, s-1-formyl-2-pyrrolidinecarboxylic acid, s-1-formylpyrrolidine-2-carboxylic acid
PubChem CID 12461843
IUPAC Name (2S)-1-formylpyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C=O)C(=O)O

Spécification

Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Specific Rotation -125
Formula Weight 143.14
Percent Purity 97%
Chemical Name or Material N-Formyl-L-proline
compliance-icons
Identifiant produit
  • N-Formyl-L-proline
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.