missing translation for 'onlineSavingsMsg'
Learn More

N-(gamma-Maleimidobutyryloxy)succinimide

Product Code. 15414349
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414349 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15414349 Supplier Thermo Scientific Alfa Aesar Supplier No. J61850.MC
 Restricted Item
Estimated Shipment: 18-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 80307-12-6 | C12H12N2O6 | 280.24 g/mol

A heterobifunctional cross-linking reagent with amine and sulfhydryl reactivity. Useful in enzyme immunoassays

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A heterobifunctional cross-linking reagent with amine and sulfhydryl reactivity
  • Useful in enzyme immunoassays
TRUSTED_SUSTAINABILITY

Specifica

CAS 80307-12-6
Melting Point 130°C to 132°C
Color White
Molecular Formula C12H12N2O6
MDL Number MFCD00036817
Quantity 100 mg
Beilstein 6427689
Synonym gmbs, 4-maleimidobutyric acid n-hydroxysuccinimide ester, 4-maleimidobutyric acid n-succinimidyl ester, n-gamma-maleimidobutyryloxy succinimide, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl butanoate, n-4-maleimidobutyryloxy succinimide, n-succinimidyl 4-maleimidobutyrate, n-succinimidyl-4-maleimidobutyrate, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxopyrrol-1-yl butanoate
InChI Key PVGATNRYUYNBHO-UHFFFAOYSA-N
SMILES O=C(CCCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-(2,5-dioxopyrrol-1-yl)butanoate
Molecular Weight (g/mol) 280.24
PubChem CID 133440
Formula Weight 280.23
Physical Form Powder
Chemical Name or Material N-(gamma-Maleimidobutyryloxy)succinimide
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.