Product Code. 10182951
Shop All Thermo Scientific Chemicals Products
Click to view available options
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
10182951
10 g
Glass bottle
10g
10602952
1 g
Glass bottle
1g
10086053
50 g
Glass bottle
50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
10182951
Supplier
Thermo Scientific Acros
Supplier No.
352770100
Estimated Shipment: 17-12-2026Log in to see stock.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
CAS: 629-94-7 | C21 H44 | 296.58 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
629-94-7
Molecular Formula
C21 H44
Molecular Weight (g/mol)
296.58
MDL Number
MFCD00009346
InChI Key
FNAZRRHPUDJQCJ-UHFFFAOYSA-N
Synonym
heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj
PubChem CID
12403
ChEBI
CHEBI:32931
IUPAC Name
henicosane
SMILES
CCCCCCCCCCCCCCCCCCCCC
Specifications
CAS Min %
97.5
CAS Max %
100.0
Melting Point
39°C to 43°C
Color
White
Boiling Point
100°C (2.0 mmHg)
Flash Point
>110°C
Infrared Spectrum
Authentic
Assay Percent Range
97.5% min. (GC)
Packaging
Glass bottle
Linear Formula
CH3 (CH2 )1 9 CH3
Quantity
10 g
Solubility Information
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,,soluble in petroleum ether
Formula Weight
296.58
Percent Purity
98%
Physical Form
Crystalline Chunks, Crystals, or Flakes
Chemical Name or Material
n-Heneicosane
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?