missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, 95%

Product Code. 15446887
Change view
Click to view available options
Quantity:
1000 mL
2500 mL
Unit Size:
1000mL
2500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446887 2500 mL 2500mL
15456887 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15446887 Supplier Thermo Scientific Alfa Aesar Supplier No. H33987.0F
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 142-82-5 | C7H16 | 100.21 g/mol

n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

Solubility
Miscible with alcohol, chloroform, and ether. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area away from incompatible substances.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -91°C
Density 0.684
Boiling Point 97°C to 99°C
Flash Point −4°C (25°F)
Odor Petroleum-like
Linear Formula CH3(CH2)5CH3
Refractive Index 1.3877
Quantity 2500 mL
UN Number UN1206
Beilstein 1730763
Merck Index 14,4659
Solubility Information Miscible with alcohol,chloroform,and ether. Insoluble in water.
Formula Weight 100.21
Percent Purity 95%
Chemical Name or Material n-Heptane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Heptane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: n-Heptane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.