missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane-d{16}, 98% (Isotopic)

Product Code. 15431807
Change view
Click to view available options
Quantity:
1 g
Packaging:
Prescored Ampule
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15431807 1 g Prescored Ampule 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15431807 Supplier Thermo Scientific Alfa Aesar Supplier No. 042289.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 33838-52-7 | C7H16 | 100.21 g/mol

n-Heptane-d{16} is used as a test fuel component in anti-knock test engines, a 100% heptane fuel is the zero point of the octane rating scale. Heptane is commercially available as mixed isomers for use in paints and coatings, as the rubber cement solvent, n-heptane for research and development and pharmaceutical manufacturing and as a minor component of gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Density 0.794 g/mL
Flash Point −4°C (24°F)
Packaging Prescored Ampule
Refractive Index 1.3877
Sensitivity Hygroscopic
Quantity 1 g
CAS 33838-52-7
Melting Point -91°C to -89°C
Boiling Point 97°C to 99°C
Molecular Formula C7H16
MDL Number MFCD00062744
UN Number UN1206
Beilstein 1730763
Synonym heptane-d16, n-heptane-d 16, 2h16 heptane, hexadecadeuterio-heptane, 2 h?? heptane, heptane-d16, 99 atom % d
Solubility Information Difficult to mix.
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-hexadecadeuterioheptane
Molecular Weight (g/mol) 100.21
PubChem CID 524600
Formula Weight 116.33
Percent Purity 98%
Chemical Name or Material n-Heptane-d{16}, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.