missing translation for 'onlineSavingsMsg'
Learn More

N-Heptane, Extra Pure, SLR, Fisher Chemical™

Product Code. 10417640 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic Container
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
500mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10417640 5 L HDPE Bottle 5 L
10417450 500 mL Amber Glass Bottle 500mL
12606717 1 L Amber Glass Bottle 1 L
10234530 2.5 L Amber Glass Bottle 2.5 L
12616717 2.5 L Plastic-coated Glass Bottle 2.5 L
10232790 5 L Plastic Container 5 L
10365750 25 L Metal Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10417640 Supplier Fisher Chemical Supplier No. H/0155/21
  • ONLINE EXCLUSIVE PRICE
    192.00 €
    190.00 € / 5 L
    Save 2.00 €
    1% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -91°C
Color Colorless
Quantity 5 L
Packaging HDPE Bottle
Physical Form Liquid
Vapor Pressure 48mbar at 20°C
Viscosity 0.4 mPaS at 20°C
Boiling Point 98°C
Formula Weight 100.2g/mol
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Heptane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.