missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, Puriss. p.a., Reagent European Pharmacopoeia,Solstice
BIOPHARMA

Product Code. 16645903
Change view
Click to view available options
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16645903 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 16645903 Supplier Burdick & Jackson BioSyn Supplier No. 65796200L
 Restricted Item
Estimated Shipment: 02-03-2027
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Specifications

Melting Point -90.5°C
CAS 142-82-5
Color Colorless
CAS Max % 1
Density 0.69 g/cm3 (15°C)
Quantity 200 L
Flash Point -4°C
Molecular Formula C7H16
Linear Formula CH3(CH2)5CH3
MDL Number MFCD00009544
Boiling Point 98.2°C to 98.4°C (1000 hPa)
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.21
Grade Pharmacopeia
Chemical Name or Material n-Heptane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.